Learn More
Penicillin-Streptomycin-Amphotericin B, 100X, MP Biomedicals
Triple antibiotic solution Concentration: Penicillin-10,000 IU/mL, Streptomycin-10mg/mL, Amphotericin B-25μg/mL Effective against yeast, fungi, gram-positive and gram-negative bacteria
Brand: MP Biomedicals 091674049
61.34 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description

Specifications
Penicillin-Streptomycin-Amphotericin B | |
Penicillin/Streptomycin/Amphotericin B | |
100 mL | |
100X Pen-Strep-Amphotericin B |
Safety and Handling
- Penicillin-Streptomycin-Amphotericin B (10,000 IU/ml, 10mg/ml, 25
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : streptomycin sulphate, penicillin G sodium salt
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.