missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentacene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.
Solubility
Soluble in organic solvents.
Notes
Air and light sensitive. Store at cool temperature.
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.
Solubility
Soluble in organic solvents.
Notes
Air and light sensitive. Store at cool temperature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 372°C to 374°C (sublimation) |
Quantity | 250 mg |
Beilstein | 1912418 |
Sensitivity | Air and light sensitive |
Merck Index | 14,7107 |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 278.35 |
Percent Purity | 97% |
Chemical Name or Material | Pentacene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'