missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentacene, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 135-48-8 |
---|---|
Molecular Formula | C22H14 |
Molecular Weight (g/mol) | 278.35 |
MDL Number | MFCD00003710 |
InChI Key | SLIUAWYAILUBJU-UHFFFAOYSA-N |
Synonym | benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene |
PubChem CID | 8671 |
ChEBI | CHEBI:33148 |
IUPAC Name | pentacene |
SMILES | C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
135-48-8 | |
278.35 | |
SLIUAWYAILUBJU-UHFFFAOYSA-N | |
8671 | |
pentacene |
C22H14 | |
MFCD00003710 | |
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
CHEBI:33148 | |
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
Specifications
135-48-8 | |
Black or Blue-Purple | |
97.5% min | |
MFCD00003710 | |
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
SLIUAWYAILUBJU-UHFFFAOYSA-N | |
pentacene | |
8671 | |
278.34 | |
Fine Crystalline Powder |
>300°C | |
Authentic | |
C22H14 | |
15, 7216 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in benzene,soluble in nitrobenzene | |
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 | |
278.35 | |
CHEBI:33148 | |
98% | |
Pentacene, 98% |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 205-193-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only