Learn More
Pentaerythritol, 98+%, Thermo Scientific Chemicals
£26.70 - £97.90
Chemical Identifiers
CAS | 115-77-5 |
---|---|
Molecular Formula | C5H12O4 |
Molecular Weight (g/mol) | 136.147 |
MDL Number | MFCD00004692 |
InChI Key | WXZMFSXDPGVJKK-UHFFFAOYSA-N |
Synonym | pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek |
PubChem CID | 8285 |
IUPAC Name | 2,2-bis(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)CO)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448783
|
Thermo Scientific Alfa Aesar
A10520.30 |
250 g |
£26.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438783
|
Thermo Scientific Alfa Aesar
A10520.0B |
1000 g |
£28.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458783
|
Thermo Scientific Alfa Aesar
A10520.0I |
5000 g |
£97.90
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.
Solubility
Soluble in Water: 0.1 g/mL
Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
Chemical Identifiers
115-77-5 | |
136.147 | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
8285 | |
C(C(CO)(CO)CO)O |
C5H12O4 | |
MFCD00004692 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
2,2-bis(hydroxymethyl)propane-1,3-diol |
Specifications
115-77-5 | |
1.396 | |
240°C (464°F) | |
MFCD00004692 | |
Hygroscopic | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
2,2-bis(hydroxymethyl)propane-1,3-diol | |
8285 | |
≥98% |
260°C to 264°C | |
276°C (30 mmHg) | |
C5H12O4 | |
1679274 | |
14,7111 | |
Soluble in Water: 0.1g/mL | |
C(C(CO)(CO)CO)O | |
136.147 | |
136.15 | |
Pentaerythritol |
Safety and Handling
H500
missing translation for 'einecsNumber' : 204-104-9
missing translation for 'rtecsNumber' : RZ2490000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only