missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 15472122
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 602-94-8
Molecular Formula C7HF5O2
Molecular Weight (g/mol) 212.08
MDL Number MFCD00002406
InChI Key YZERDTREOUSUHF-UHFFFAOYSA-N
Synonym pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
PubChem CID 11770
ChEBI CHEBI:46796
IUPAC Name 2,3,4,5,6-pentafluorobenzoic acid
SMILES OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 100.0°C to 104.0°C
Boiling Point 220.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5CO2H
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot water
Formula Weight 212.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzoic acid
compliance-icons
Product Identifier
  • Pentafluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'