Learn More
Pentafluorophenol, 99%, Thermo Scientific Chemicals
£22.90 - £203.00
Chemical Identifiers
CAS | 771-61-9 |
---|---|
Molecular Formula | C6HF5O |
Molecular Weight (g/mol) | 184.07 |
MDL Number | MFCD00002156 |
InChI Key | XBNGYFFABRKICK-UHFFFAOYSA-N |
Synonym | pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp |
PubChem CID | 13041 |
IUPAC Name | 2,3,4,5,6-pentafluorophenol |
SMILES | OC1=C(F)C(F)=C(F)C(F)=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453047
|
Thermo Scientific Alfa Aesar
A15574.06 |
5 g |
£22.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463047
|
Thermo Scientific Alfa Aesar
A15574.14 |
25 g |
£72.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473047
|
Thermo Scientific Alfa Aesar
A15574.22 |
100 g |
£203.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentafluorophenol is used in the preparation of pentafluorophenyl esters for peptide synthesis and vulcanized polymers. It is also used for peptide coupling and heterocyclic acid derivatives. It is involved in the preparation of pentafluorophenyl formate which acts as a formulating agent for amines and amino acids.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, bases, acid chlorides and acid anhydrides.
Chemical Identifiers
771-61-9 | |
184.07 | |
XBNGYFFABRKICK-UHFFFAOYSA-N | |
13041 | |
OC1=C(F)C(F)=C(F)C(F)=C1F |
C6HF5O | |
MFCD00002156 | |
pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp | |
2,3,4,5,6-pentafluorophenol |
Specifications
771-61-9 | |
1.757 | |
72°C (161°F) | |
1.427 | |
1912584 | |
Soluble in water. | |
OC1=C(F)C(F)=C(F)C(F)=C1F | |
184.07 | |
184.06 | |
Pentafluorophenol |
32°C to 36°C | |
143°C | |
C6HF5O | |
MFCD00002156 | |
pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp | |
XBNGYFFABRKICK-UHFFFAOYSA-N | |
2,3,4,5,6-pentafluorophenol | |
13041 | |
99% |
Sicherheit und Handhabung
GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
H302+H312+H332-H314-H318-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 212-235-8
missing translation for 'rtecsNumber' : SM6680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only