missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-morpholinobenzoate, 97%, Thermo Scientific™

Product Code. 10183674
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-59-8
Molecular Formula C17H12F5NO3
Molecular Weight (g/mol) 373.28
MDL Number MFCD09025824
InChI Key SMSUNMKSTOOWTF-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholinobenzoate, pentafluorophenyl 2-morpholin-4-ylbenzoate, pentafluorophenyl 2-morpholin-4-yl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl benzoate, perfluorophenyl 2-morpholinobenzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-ylbenzoate, 2-4-morpholinyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229476
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-ylbenzoate
Formula Weight 373.28
Percent Purity 95%
Chemical Name or Material Pentafluorophenyl 2-morpholinobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.