missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-(morpholinosulfonyl)benzoate, 97%, Thermo Scientific™

Product Code. 10769215
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-27-7
Molecular Formula C17H12F5NO5S
Molecular Weight (g/mol) 437.34
MDL Number MFCD09879986
InChI Key FDXYCKHOIXBDOA-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholinosulfonyl benzoate, pentafluorophenyl 2-morpholine-4-sulfonyl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholine-4-sulfonyl benzoate, pentafluorophenyl 2-morpholin-4-ylsulphonyl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholinosulfonylbenzoate, 2,3,4,5,6-pentafluorophenyl 2-4-morpholinylsulfonyl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-ylsulfonylbenzoate, 2-4-morpholinylsulfonyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229773
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-ylsulfonylbenzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2S(=O)(=O)N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 437.34
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-(morpholinosulfonyl)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.