missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Thermo Scientific™

Product Code. 11360587
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Identificatori chimici

CAS 915707-42-5
Molecular Formula C17H9F5N2O2
Molecular Weight (g/mol) 368.26
MDL Number MFCD09702352
InChI Key LGHZGIHYFYLEAC-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, perfluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1-methylpyrazol-3-yl benzoate, 4-1-methyl-3-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229473
SMILES CN1C=CC(=N1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifica

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2,3,4,5,6-pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
Formula Weight 368.26
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'