missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate, 97%, Thermo Scientific™

Product Code. 10366754
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 941717-00-6
Molecular Formula C18H11F5N2O2
Molecular Weight (g/mol) 382.29
MDL Number MFCD09879978
InChI Key GQQALUKKLLEHOR-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-3,5-dimethyl-1h-pyrazol-1-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-3,5-dimethylpyrazol-1-yl benzoate, pentafluorophenyl 4-3,5-dimethylpyrazol-1-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-3,5-dimethylpyrazol-1-yl benzoate, 4-3,5-dimethyl-1-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229762
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-(3,5-dimethylpyrazol-1-yl)benzoate
SMILES CC1=CC(=NN1C2=CC=C(C=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 382.29
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-(3, 5-dimethyl-1H-pyrazol-1-yl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'