missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-morpholinobenzoate, 97%, Thermo Scientific™

Product Code. 10537634
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-51-4
Molecular Formula C17H12F5NO3
Molecular Weight (g/mol) 373.28
MDL Number MFCD09817462
InChI Key UJYQDNSLPIRXRT-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-morpholin-4-ylbenzoate, pentafluorophenyl 4-morpholinobenzoate, pentafluorophenyl 4-morpholin-4-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-morpholin-4-yl benzoate, perfluorophenyl 4-morpholinobenzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-morpholin-4-ylbenzoate, 4-4-morpholinyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229462
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-morpholin-4-ylbenzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=C(C=C2)N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 373.28
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-morpholinobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'