missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 6-phenylnicotinate, 97%, Thermo Scientific™

£35.70 - £80.50

Chemical Identifiers

CAS 934570-41-9
Molecular Formula C18H8F5NO2
Molecular Weight (g/mol) 365.26
MDL Number MFCD09702345
InChI Key YUPUNKGUYBKZST-UHFFFAOYSA-N
Synonym pentafluorophenyl 6-phenylnicotinate, pentafluorophenyl 2-phenylpyridine-5-carboxylate, pentafluorophenyl 6-phenylpyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-phenylpyridine-3-carboxylate, 6-phenyl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229464
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 6-phenylpyridine-3-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CC=C2)C(F)=C1F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10632295
View Documents
Thermo Scientific Maybridge
CC21126CB
250 mg Amber Glass Bottle
£35.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10545364
View Documents
Thermo Scientific Maybridge
CC21126DA
1 g Amber Glass Bottle
£80.50
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

934570-41-9
365.26
YUPUNKGUYBKZST-UHFFFAOYSA-N
24229464
FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CC=C2)C(F)=C1F
C18H8F5NO2
MFCD09702345
pentafluorophenyl 6-phenylnicotinate, pentafluorophenyl 2-phenylpyridine-5-carboxylate, pentafluorophenyl 6-phenylpyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-phenylpyridine-3-carboxylate, 6-phenyl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) 6-phenylpyridine-3-carboxylate

Specifications

934570-41-9
100.0
C18H8F5NO2
pentafluorophenyl 6-phenylnicotinate, pentafluorophenyl 2-phenylpyridine-5-carboxylate, pentafluorophenyl 6-phenylpyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-phenylpyridine-3-carboxylate, 6-phenyl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CC=C2)C(F)=C1F
365.26
365.26
Pentafluorophenyl 6-phenylnicotinate
96.5
97%
MFCD09702345
YUPUNKGUYBKZST-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 6-phenylpyridine-3-carboxylate
24229464
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Certificates