missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentan-1-ol, Extra Pure, SLR, Fisher Chemical™
Chemical Identifiers
CAS | 71-41-0 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00002977,MFCD00081734,MFCD01075169 |
InChI Key | AMQJEAYHLZJPGS-UHFFFAOYSA-N |
Synonym | 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol |
PubChem CID | 6276 |
ChEBI | CHEBI:44884 |
IUPAC Name | pentan-1-ol |
SMILES | CCCCCO |
Description
Pentan-1-ol, Extra Pure, SLR
Chemical Identifiers
71-41-0 | |
88.15 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
6276 | |
pentan-1-ol |
C5H12O | |
MFCD00002977,MFCD00081734,MFCD01075169 | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CHEBI:44884 | |
CCCCCO |
Specifications
71-41-0 | |
Colorless | |
137°C | |
C5H12O | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CCCCCO | |
88.15 | |
3 | |
88.15g/mol |
-78°C | |
7 | |
Plastic Coated Glass bottle | |
MFCD00002977,MFCD00081734,MFCD01075169 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
pentan-1-ol | |
6276 | |
CHEBI:44884 | |
Liquid |