missing translation for 'onlineSavingsMsg'
Learn More
Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™ Product Code.: 15664720

Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15664720
2.5 L
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Repeated exposure may cause skin dryness or cracking.
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15664720

Brand: Honeywell-Chromasolv 349562.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Repeated exposure may cause skin dryness or cracking.
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for HPLC,≥99.0%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Density 0.626g/mL at 25°C
Boiling Point 35°C to 36°C
Quantity 2.5 L
Refractive Index n20/D 1.358
Linear Formula CH3(CH2)3CH3
Beilstein 969132
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
Formula Weight 72.15g/mol
Percent Purity ≥99%
Grade CHROMASOLV™
Chemical Name or Material Pentane, for HPLC
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.