missing translation for 'onlineSavingsMsg'
Learn More

Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Honeywell Riedel-de Haën™

Product Code. 15687040
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons),≥99.0%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Density 0.626g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Formula Weight 72.15g/mol
Percent Purity ≥99%, ≥99% (GC)
Grade CHROMASOLV™
Chemical Name or Material Pentane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'