missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Honeywell Riedel-de Haën™
CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons),≥99.0%
£96.02 - £174.54
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
109-66-0 | |
0.083 | |
500°F | |
CH3(CH2)3CH3 | |
UN1265 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
≥99%, ≥99% (GC) | |
Pentane |
0.626g/mL at 25°C | |
−49°C (−56.2°F) | |
C5H12 | |
MFCD00009498 | |
969132 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15g/mol | |
CHROMASOLV™ |
Safety and Handling
P210-P261-P273-P301 + P310-P331
missing translation for 'rtecsNumber' : RZ9450000