Learn More
Thermo Scientific™ pENTR™/D-TOPO™ Cloning Kit
Utilize a highly efficient, 5-minute cloning strategy (TOPO™ Cloning) to directionally clone a blunt-end PCR product into a vector for entry into the Gateway™ System or the MultiSite Gateway™ System
Brand: Thermo Scientific™ K240020SP
332.01 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.40000kg
Description
- The pENTR™/D-TOPO™ Cloning Kits utilize a highly efficient, 5-minute cloning strategy (TOPO™ Cloning) to directionally clone a blunt-end PCR product into a vector for entry into the Gateway™ System or the MultiSite Gateway™ System. Blunt-end PCR products clone directionally at greater than 90% efficiency, with no ligase, post-PCR procedures, or restriction enzymes required.
- Gateway™ System Ready—Rapidly shuttle cloned genes between multiple vector systems
- Fast and Easy—Go from PCR to Gateway™ Entry clone in just 3 steps and as little as ∼5 minutes hands on time
- Efficient—Achieve over 90% clones with correct insert in the right direction
- Proven—Reliable performance for over a decade
- For access to the Gateway™ System, just PCR amplify your gene of interest and add the product straight to the provided topoisomerase charged pENTR™/D-TOPO™ vector, incubate 5 minutes, and transform the provided competent E. coli cells. The resulting attL containing Gateway™ Entry clones are ready for efficient recombination with your choice of Gateway™ Destination vectors
- The pENTR™/D-TOPO™ vector (Figure 1) includes M13 and T7 primer sequencing sites and attL recombination sites flanking the PCR product insertion site. This allows clones to be easily sequence verified and recombined into your choice of attR containing Gateway™ destination vectors. A Kanamycin resistance gene and a pUC origin are used for selection and high-copy propagation in E. coli.
- With Directional TOPO™ cloning technology there is no need for PCR clean up, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase charged vector, incubate 5 minutes, transform, and obtain up to 90% directionally inserted clones. A four base overhang on the vector pairs with a four base sequence designed into the forward primer used in your PCR reaction to provide directionality to the topoisomerase ligation reaction (Figure 2).
- Gateway™ recombination cloning technology circumvents the limitations of restriction mediated cloning, enabling you to access virtually any expression system in a simple one hour, 99%-efficient and reversible, Gateway™ recombination reaction. The ability to move the same sequence of DNA between different vectors without using restriction enzymes, ligase, subcloning steps, screening of countless colonies, or re-sequencing will help save you time, money, and effort.
- When it comes to cloning, TOPO™ cloning technology and Gateway™ recombination cloning technology have been a reliable partner for thousands of scientists for over ten years. Fast, simple to use, and efficient, TOPO™ cloning and Gateway™ recombina
Specifications
TOP10 | |
TOPO cloning | |
Kit | |
20 Reactions | |
T7 |
Chemically Competent E. coli | |
pENTR™/D-TOPO™ Cloning Kit contains pENTR/D-TOPO™ vector, dNTPs, salt solution, sterile water, and universal M13 sequencing primers. Store components at -20°C. |
|
Cloning | |
Cloning Kit |