missing translation for 'onlineSavingsMsg'
Learn More

Pentraxin 2/SAP Antibody (APCS/3240), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79848B

Product Code. 18253018

  • £470.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Pentraxin 2/SAP
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
APCS
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) (Uniprot: P02743)
0.1 mL
Neuroscience
Human
IgG2b κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
APCS/3240
Immunohistochemistry, Immunohistochemistry-Paraffin
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers