missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentraxin 2/SAP Antibody (APCS/3240), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79848PCP
This item is not returnable.
View return policy
Description
Pentraxin 2/SAP Monoclonal antibody specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Pentraxin 2/SAP | |
Monoclonal | |
PerCP | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component | |
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) (Uniprot: P02743) | |
0.1 mL | |
Neuroscience | |
325 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
APCS/3240 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |