missing translation for 'onlineSavingsMsg'
Learn More

Pentraxin 2/SAP, Mouse anti-Human, Clone: APCS/3240, Novus Biologicals™

Mouse Monoclonal Antibody

£206.00 - £375.00

Specifications

Antigen Pentraxin 2/SAP
Clone APCS/3240
Concentration 0.2 mg/mL
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18265607
View Documents Promotion Details
Novus Biologicals
NBP2-79714-20UG
20 μg
£206.00
20µg
Please to purchase this item. Need a web account? Register with us today!
18221068
View Documents Promotion Details
Novus Biologicals
NBP2-79714-100UG
100 μg
£375.00
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Pentraxin 2/SAP
0.2 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Human
P02743, P02743
325
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary)
Primary
Store at 4°C.
APCS/3240
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component
APCS
IgG2b κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications