Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
Brand: Thermo Scientific™ 174501
Additional Details : Weight : 0.35000kg
Description
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
Specifications
30 μm | |
Two PEEK Tubes with nanoViper™ Fittings | |
22,000 psi (1500 bar) | |
PepMap Neo | |
PEEK Tubing with nanoViper™ Fittings |
PepMap™ Neo Trap Cartridge (part number 174500) | |
100 mm | |
60°C | |
PEEK | |
Trap and Elute, Peptide Mapping |