Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples.
Brand: Thermo Scientific™ 174500
341.83 GBP valid until 2024-07-31
Use promo code "21824" to get your promotional price.
Additional Details : Weight : 0.06300kg
Descrizione
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with Double nanoViper™ PepMap™ Neo UHPLC Columns or EASY-Spray™ PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples. Connect the trap cartridge to your HPLC system with the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings (Catalog Numbers 174501 and 174502).
Specifica
Trap Cartridge | |
PepMap™ Neo Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings (part number 174502) | |
22,000 psi (1500 bar) | |
2 to 8 | |
450 m2/g | |
L1 | |
PepMap Neo | |
15% | |
Yes | |
PepMap™ Neo 5 μm C18 300 μm X 5 mm Trap Cartridge |
300 μm | |
5 mm | |
5 μm | |
100 Å | |
60°C | |
C18 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase | |
Trap and Elute |