Learn More
Pepsin, Extra Pure, Powder, SLR, Fisher Chemical™
Enzyme that breaks down proteins into smaller peptides.
Brand: Fisher Chemical P/1120/53
350.34 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 21
Additional Details : CAS Number : 9001-75-6 Weight : 0.50000kg
Description
Enzyme that breaks down proteins into smaller peptides.
Specifications
500 g |
Safety and Handling
- Pepsin
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.