missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepstatin A, Fisher BioReagents
73.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
250 mg
Packaging:
Amber Glass
Description
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 235°C |
Color | White |
Melting Range | 230° to 240°C |
Infrared Spectrum | Conforms to reference |
Packaging | Amber Glass |
Quantity | 5 mg |
Merck Index | 15, 7260 |
Formula Weight | 685.9 |
Physical Form | Powder or Solid |
Chemical Name or Material | Pepstatin A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'