missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, 0%, Honeywell™ Fluka™

Product Code. 15662820
Click to view available options
Quantity:
5 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00002128
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Density 1.13g/mL at 25°C
Linear Formula CH3CO3H
Quantity 100 mL
Beilstein 1098464
Formula Weight 76.05g/mol
Vapor Pressure 20mmHg (25°C)
Percent Purity 0%
Chemical Name or Material Peracetic Acid Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'