missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peracetic Acid Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
500 mL
Specifications
CAS | 79-21-0 |
Molecular Formula | C2H4O3 |
Linear Formula | CH3CO3H |
MDL Number | MFCD00002128 |
Quantity | 500 mL |
Beilstein | 1098464 |
Synonym | peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1 |
InChI Key | KFSLWBXXFJQRDL-UHFFFAOYSA-N |
SMILES | CC(=O)OO |
IUPAC Name | ethaneperoxoic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'