missing translation for 'onlineSavingsMsg'
Learn More
Perchloric Acid, 0.1M, EP Grade, Reagecon™ Product Code.: 11444799

Perchloric Acid, 0.1M, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11444799 Shop All Reagecon Products
1 L, Tamper evident
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
This item is not returnable. View return policy

Product Code. 11444799

Brand: Reagecon 3003900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.2.2) and prepared in an ISO 9001 facility. Perchloric Acid, 0.1M, EP Grade, Reagecon™ is a ready-to-use volumetric solution with 8.5mL/L HClO4 in acetic acid. It is a superacid used as a laboratory reagent.

Perchloric acid is a strong mineral acid that is useful in preparing perchloric salts. It functions as a powerful oxidizer when hot. This solution is prepared by mixing 8.5mL perchloric acid and about 900mL glacial acetic acid, adding 30mL acetic anhydride and then diluting to 1L with glacial acetic acid. This yields a 0.1M solution with about 10.046g/L HClO4.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.2.2) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise volumetric solutions

Specifications

Color Colorless
CAS 7601-90-3, 64-19-7
Concentration 0.1M
Odor Odorless
Packaging Tamper evident
Certificate of Analysis Yes
Solvent or Matrix Glacial acetic acid
Quantity 1 L
Molecular Formula ClHO4
Physical Form Liquid
MDL Number MFCD00011325
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46g/mol
Concentration or Composition (by Analyte or Components) 8.5mL Perchloric acid per L = 10.046g
Chemical Name or Material Perchloric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP20125A1 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog Number3003900
Lot NumberP20124K2 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number3003900
Lot NumberP20124H2 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number3003900
Lot NumberP20124E1 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number3003900
Lot NumberP20124A1 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number3003900
compliance-icons compliance-icons
Product Identifier
  • 0,1M perchloric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perchloric Acid, 0.1M, EP Grade, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.