missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 60%, ACS Reagent, Honeywell™ Fluka™
ACS Reagent, 60%
£55.22 - £265.39
Chemical Identifiers
CAS | 7601-90-3 |
---|---|
Molecular Formula | ClHO4 |
Molecular Weight (g/mol) | 100.45 |
MDL Number | MFCD00011325 |
InChI Key | VLTRZXGMWDSKGL-UHFFFAOYSA-N |
Synonym | unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 |
PubChem CID | 24247 |
ChEBI | CHEBI:29221 |
IUPAC Name | perchloric acid |
SMILES | O[Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15643270
|
Honeywell-Fluka
311413-100ML |
100 mL | Glass Bottle | N/A | |||||
15673270
|
Honeywell-Fluka
311413-500ML |
500 mL | Glass Bottle |
£156.49
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15653270
|
Honeywell-Fluka
311413-1L |
1 L | Glass Bottle |
£265.39
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
24247 | |
perchloric acid |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
CHEBI:29221 | |
O[Cl](=O)(=O)=O |
Specifications
7601-90-3 | |
203°C | |
≤0.003% | |
113°C (235.4°F) | |
HClO4 | |
UN1873 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
24247 | |
100.46g/mol | |
ACS Reagent |
-18°C | |
1.535g/mL at 25°C | |
7mmHg (25°C) | |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
O[Cl](=O)(=O)=O | |
100.45 | |
CHEBI:29221 | |
60% | |
Perchloric Acid |
Safety and Handling
P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
H271-H290-H302-H314-H373
missing translation for 'dotInformation' : 8(5.1)/PGI