Learn More
Perchloric acid, ACS, 48-50%, Thermo Scientific Chemicals
Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm
£23.61 - £259.29
Chemical Identifiers
CAS | 7601-90-3 |
---|---|
Molecular Formula | ClHO4 |
Molecular Weight (g/mol) | 100.45 |
MDL Number | MFCD00011325 |
InChI Key | VLTRZXGMWDSKGL-UHFFFAOYSA-N |
Synonym | unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 |
PubChem CID | 24247 |
ChEBI | CHEBI:29221 |
IUPAC Name | perchloric acid |
SMILES | O[Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15441997
|
Thermo Scientific Alfa Aesar
044464.AE |
100 mL | N/A | ||||||
15451997
|
Thermo Scientific Alfa Aesar
044464.AP |
500 mL |
£96.78
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15461997
|
Thermo Scientific Alfa Aesar
044464.K4 |
2.5 L |
£259.29
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
24247 | |
perchloric acid |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
CHEBI:29221 | |
O[Cl](=O)(=O)=O |
Specifications
7601-90-3 | |
1.41 g/cm3 at 20°C | |
48-50% | |
Liquid | |
UN1802 | |
Miscible with water. | |
O[Cl](=O)(=O)=O | |
100.45 | |
CHEBI:29221 | |
ACS |
°C to 112°C | |
29mm Hg at 20°C | |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
24247 | |
100.46 | |
Perchloric acid |
Safety and Handling
Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
H271-H290-H302-H314-H335-H373
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 231-512-4
missing translation for 'rtecsNumber' : SC7500000
missing translation for 'tsca' : Yes
RUO – Research Use Only