missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric acid in anhydrous acetic acid, BAKER ANALYZED™, J.T. Baker™
Click to view available options
1 L
Specifications
Specifications
CAS | 7601-90-3 |
Concentration | 0.1 mol/L (0.1 N) |
Quantity | 1 L |
Molecular Formula | ClHO4 |
MDL Number | MFCD00011325 |
InChI Key | VLTRZXGMWDSKGL-UHFFFAOYSA-N |
SMILES | O[Cl](=O)(=O)=O |
IUPAC Name | perchloric acid |
Molecular Weight (g/mol) | 100.45 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'