missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric acid concentrate, For 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium, Honeywell Fluka™
Click to view available options
Quantity:
1 L
6 x 1 Ea.
Packaging:
Ampoule
Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 203°C |
CAS Min % | ≥70.0000%, ≥20.0000% |
CAS Max % | <90.0000%, <30.0000% |
Density | 1.66 g/cm3 |
Packaging | Ampoule |
Flash Point | 113°C (235.4°F) |
Quantity | 6 x 1 Ea. |
Linear Formula | HClO4 |
Physical Form | Concentrate |
UN Number | UN1873 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'