missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 348630025


 View more versions of this product

Product Code. 10043213

  • £137.69 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Perchloric acid
0.1 M Solution in Acetic Acid
1.0600g/mL
1.06
2.5 L
Liquid
2920
15, 7267
Solubility in water: insoluble.
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
Pure
Colorless
7601-90-3, 64-19-7
Plastic Bottle
40°C
ClHO4
MFCD00011325
01,796; 02,309; 03,220; 06,453; 09,279; 11,402
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Perchloric acid, 0.1 M solution in acetic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only