missing translation for 'onlineSavingsMsg'
Learn More
Perchloric Acid Solution, 1M, Fisher Chemical™ Product Code.: 12993564

Perchloric Acid Solution, 1M, Fisher Chemical™
BIOPHARMA

Product Code. 12993564
1000 mL, 1000mL
Click to view available options
Quantity:
1000 mL
Packaging:
1000mL
This item is not returnable. View return policy

Product Code. 12993564

Brand: Fisher Chemical VL 252 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 1mol/L
Packaging 1000mL
Quantity 1000 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1096375 Certificate TypeCertificate of Analysis Date22/03/2025 Catalog NumberVL 252 1000
Lot Number1092923 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog NumberVL 252 1000
Lot Number1077648 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 252 1000
Lot Number1071532 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 252 1000
Lot Number1005560 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 252 1000
compliance-icons compliance-icons
Product Identifier
  • Perchloric acid 1M Volumetric Solution (VL 252)
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : perchloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perchloric Acid Solution, 1M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.