missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Poly Bottle
Beschreibung
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions

Spezifikation
Spezifikation
Color | Colorless |
Melting Point | -18°C |
Boiling Point | 203°C |
Density | 1.66g/cu.cm |
Packaging | Poly Bottle |
Flash Point | 113°C (235.4°F) |
Quantity | 1 L |
Linear Formula | HClO4 |
Physical Form | Liquid |
UN Number | UN1873 |
Mehr anzeigen |
Sicherheit und Handhabung




Produktidentifikator
- Perchloric acid
- Danger
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H271-May cause fire or explosion; strong oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Perchloric acid
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Perchloric Acid, TraceSELECT™ Ultra, 67 to 72%, Honeywell Fluka™ >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur