missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ Centrifugation Media

For separation of cells, subcellular particles, and larger viruses (down to approx. 70S) under gentle conditions that preserve viability and morphological integrity. Cytiva™ Percoll™ Centrifugation Media Percoll is composed of colloidal silica coated with polyvinylpyrrolidone (PVP).

£147.17 - £351.00

Specifications

Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 15cP
Osmolality < 25 mOsm/kg
Color Colorless to Slightly Yellow
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11530734
View Documents
Cytiva
17-0891-02
250 mL
£147.17
250mL
In Stock
to see stock.
Edge
Add to basket
 
10607095
View Documents Promotion Details
Cytiva
17-0891-01
1 L
£351.00
1 L
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Percoll is a silica sol with nondialyzable PVP coating; 15-30nm dia. bead size

For in vitro research use only.

Specifications

Specifications

4–30°C
15cP
Colorless to Slightly Yellow
Centrifugation Media
Liquid
Cell Isolation
< 25 mOsm/kg
8.5 to 9.5
1.135 g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 years

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.