missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Percoll™, MP Biomedicals™
Click to view available options
Quantity:
100 mL
Description
Centrifugation of Percoll™ results in spontaneous formation of a density gradient due to the heterogeneity of particle sizes in the medium. Percoll™ can be used for formation of gradients either by the use of conventional gradient mixers or by high speed centrifugation. In the latter case the sample can be premixed with the medium and then separated on the gradient created in situ. In this way gradient formation and sample separation can be achieved in a single operation.
Specifications
Specifications
CAS | 65455-52-9 |
Density | 1.128g/mL (Lit.) |
Quantity | 100 mL |
Viscosity | Record cp at 20°C |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction