missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ PLUS Centrifugation Media

Low endotoxin reagent for density gradient centrifugation of cells, viruses, and subcellular particles. Cytiva™ Percoll™ PLUS Centrifugation Media is non-toxic, shows low viscosity and is ideal for density gradients.

Brand:  Cytiva 17-5445-02

 View more versions of this product

Product Code. 11500744

  • £202.80 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Silica sol coated with covalently linked silane; 15-30nm dia. bead size
  • Low endotoxin levels (max. 2 EU/mL); well-suited for cell separation in clinical research applications

For in vitro research use only.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
Cell Isolation
2 EU/mL
Colorless to Slightly Yellow
Centrifugation Media
Liquid
4–30°C
15cP
< 30 mOsm/kg
8.9 to 9.9
1.135 g/mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-1.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 1.5% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Percoll™ PLUS Centrifugation Media > Percoll PLUS; Silane-coated; 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.