missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
£165.00 - £768.12
Specifications
Storage | Room Temperature |
---|---|
For Use With | GC/MS |
Standard Concentration | 98 to 100% |
Product Code | Brand | Standard Components | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Components | Quantity | Price | Quantity & Availability | ||||
11567740
|
Thermo Scientific™
TS-63163 |
Heptafluoro-butanoic acid anhydride | 25 g |
£213.69
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11772247
|
Thermo Scientific™
TS-63164 |
Heptafluoro-butanoic acid anhydride | 10 x 1 mL |
£236.00
Pack of 10 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11587740
|
Thermo Scientific™
TS-65191 |
Pentafluoro-propanoic acid anhydride | 100 g |
£623.36
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11507750
|
Thermo Scientific™
TS-65193 |
Pentafluoro-propanoic acid anhydride | 10 x 1 mL |
£768.12
10mL |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11517800
|
Thermo Scientific™
TS-67363 |
Trifluoroacetic acid anhydride | 100 g |
£432.31
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11557740
|
Thermo Scientific™
TS-63162 |
Heptafluoro-butanoic acid anhydride | 100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11597740
|
Thermo Scientific™
TS-65192 |
Pentafluoro-propanoic acid anhydride | 25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
- Used to prepare fluoracyl derivatives for GC/MS
- Produce stable volatile derivatives for FID and ECD techniques
Specifications
Room Temperature | |
GC/MS | |
98 to 100% |
For Research Use Only. Not for use in diagnostic procedures.