missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
£29.85 - £242.00
Chemical Identifiers
CAS | 311-89-7 |
---|---|
Molecular Formula | C12F27N |
Molecular Weight (g/mol) | 670 |
MDL Number | MFCD00000436 |
InChI Key | RVZRBWKZFJCCIB-UHFFFAOYSA-N |
Synonym | perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine |
PubChem CID | 9397 |
ChEBI | CHEBI:38854 |
IUPAC Name | 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine |
SMILES | C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447320
|
Thermo Scientific Acros
123770100 |
10 g | Glass bottle | N/A | |||||
10081160
|
Thermo Scientific Acros
123771000 |
100 g | Glass bottle | N/A | |||||
10091160
|
Thermo Scientific Acros
123772500 |
250 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
311-89-7 | |
670 | |
RVZRBWKZFJCCIB-UHFFFAOYSA-N | |
9397 | |
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine |
C12F27N | |
MFCD00000436 | |
perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine | |
CHEBI:38854 | |
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Specifications
311-89-7 | |
Colorless | |
173.0°C to 181.0°C | |
C12F27N | |
MFCD00000436 | |
perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine | |
RVZRBWKZFJCCIB-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine | |
9397 | |
2.3-3.3 mm2/s (25°C) | |
Liquid |
-52.0°C | |
1.8800g/mL | |
Authentic | |
1.291 | |
1.88 | |
Solubility in water: insoluble. | |
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
670 | |
CHEBI:38854 | |
670 | |
Perfluoro-compound FC-43* |
Safety and Handling
missing translation for 'einecsNumber' : 206-223-1
missing translation for 'rtecsNumber' : YA1000000
missing translation for 'tsca' : TSCA
Product Identifier
- Perfluoro-compound FC-43]TS
RUO – Research Use Only