missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro(methylcyclohexane), 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for fluorous biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for ″fluorous″ biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.
Solubility
Miscible with acetone and benzene.
Notes
Incompatible with strong oxidizing agents.
Perfluoro(methylcyclohexane) is used as a solvent to determine fluorophilicity of hydrocarbon and fluorocarbon-functionalized nicotinic acid esters. It combines with chloroform and acts as an ingredient for ″fluorous″ biphase reactions. Furthermore, it finds application as a heat transfer agent, a dielectric fluid and a perfluorocarbon tracer. It acts as a good solvent for gases.
Solubility
Miscible with acetone and benzene.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -70°C |
Density | 1.8 |
Boiling Point | 75°C to 76°C |
Odor | Odorless |
Refractive Index | 1.3 00 |
Quantity | 25 g |
Beilstein | 1915981 |
Solubility Information | Miscible with acetone and benzene. |
Formula Weight | 350.06 |
Percent Purity | 94% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'