missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorooctanoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Perfluorooctanoyl chloride is derivatizated and used in the determination of Amphetamine and Methamphetamine in Blood. It can be used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents and moisture. Store under dry inert gas. Incompatible with strong oxidizing agents, strong bases, alcohols.
Perfluorooctanoyl chloride is derivatizated and used in the determination of Amphetamine and Methamphetamine in Blood. It can be used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents and moisture. Store under dry inert gas. Incompatible with strong oxidizing agents, strong bases, alcohols.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.744 |
Boiling Point | 130°C to 132°C |
Refractive Index | 1.3045 |
Quantity | 5 g |
UN Number | UN3265 |
Beilstein | 1809677 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoyl chloride |
Formula Weight | 432.52 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'