Learn More
Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Description
Perfluorotributylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as a solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Perfluorotributylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as a solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.
Solubility
Miscible with chloroform, cyclohexane, ethyl ether, ethyl acetate, heptane, carbon tetrachloride and petroleum ether. Immiscible with water, methanol, toluene, isopropyl alcohol, benzene and acetone.
Notes
Store in a cool place. Incompatible with powdered metals and alkali metals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -52°C |
Density | 1.88 |
Boiling Point | 177°C to 180°C |
Refractive Index | 1.289 |
Quantity | 1 g |
Beilstein | 1813883 |
Solubility Information | Miscible with chloroform,cyclohexane,ethyl ether,ethyl acetate,heptane,carbon tetrachloride and petroleum ether. Immiscible with water,methanol,toluene,isopropyl alcohol,benzene and acetone. |
Formula Weight | 671.1 |
Chemical Name or Material | Perfluorotributylamine, Mass Spec Standard |
RUO – Research Use Only