Learn More
BINDER™ Perforated Shelf Rack
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 130, KBF/KBF PRO 130
KB/KB PRO 720, KBF/KBF PRO 720
KB/KB PRO/KBF/KBF PRO 260/470
KBF/KBF PRO 1600, KB PRO 1600
Unit Size:
Each
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
Specifications
| For Use With (Equipment) | KB/KB PRO 1060, KBF/KBF PRO 1060 |
| Material | Stainless Steel |
| Type | Rack |