missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain AstiPure™ PFD1 Controlled Flow Pump
Click to view available options
Flow Rate:
10 L/min.
Unit Size:
Each
Description
- Pneumatically operated: two bellows made from PTFE reciprocate horizontally
- Pumping frequency is much lower than an equivalent diaphragm pump
- All AstiPure PFD pump wetted components are manufactured with High Purity PFA and PTFE
- All other components are manufactured with high technology materials, such as PVDF and PEEK, ensuring no ionic contamination, even if there is a bellows failure
- Pumps are assembled in cleanroom, tested with DI water, cleaned, then double sealed in plastic bags
- No metal parts (internal or external)
- Easy maintenance
- No rotating shafts or glands to cause leakage
- Pneumatically operated (dry air or nitrogen)
- Self-priming
- Suitable for liquid temperatures from 0°C (32°F) up to 100°C (212°F)
- Compact design
- Recirculation, dispensing, and filtration with controlled flow rates and volumes
- Chemical injection and sampling.
- Transfer of chemicals and solvents in cleanrooms
- safe transfer of corrosive chemicals and solvents in hazardous areas
- Transfer and dispensing of pharmaceuticals
Specifications
Specifications
| Flow Rate | 10 L/min. |
| Temperature (Metric) Min. | 0°C |
| Temperature (Metric) Max | 100°C |
| Maximum Pressure | 4 bar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction