missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perhexiline maleate, >99%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Carnitine palmitoyltransferase 1 and 2 (CPT1/2) inhibitor. Inhibits palmitate oxidation in rat cardiomyocytes and increases cardiac efficiency in isolated rat hearts. Antianginal.
Specifications
Specifications
Linear Formula | C19H35N.C4H4O4 |
Quantity | 10 mg |
Solubility Information | Soluble to 100mM in DMSO and to 20mM in ethanol |
Formula Weight | 393.56 |
Chemical Name or Material | 2-(2,2-Dicyclohexylethyl)piperidine maleate |
Grade | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction