missing translation for 'onlineSavingsMsg'
Learn More

Perhexiline maleate, >99%, Tocris Bioscience™

Product Code. 30001012
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Carnitine palmitoyltransferase 1 and 2 (CPT1/2) inhibitor

Carnitine palmitoyltransferase 1 and 2 (CPT1/2) inhibitor. Inhibits palmitate oxidation in rat cardiomyocytes and increases cardiac efficiency in isolated rat hearts. Antianginal.

Chemical Identifiers

CAS 6724-53-4
Molecular Formula C23H39NO4
Molecular Weight (g/mol) 393.568
InChI Key JDZOTSLZMQDFLG-BTJKTKAUSA-N
Synonym perhexiline maleate, perhexiline maleate salt, pexid, 2-2,2-dicyclohexylethyl piperidine maleate, perhexiline maleate usan, ccris 5491, rac perhexiline maleate, 2-2,2-dicyclohexylethyl piperidine acid maleate, 2-2,2-dicyclohexylethyl piperidine maleate 1:1, 2-2,2-dicyclohexylethyl piperidinium hydrogen maleate
PubChem CID 5284439
IUPAC Name (Z)-but-2-enedioic acid;2-(2,2-dicyclohexylethyl)piperidine
SMILES C1CCC(CC1)C(CC2CCCCN2)C3CCCCC3.C(=CC(=O)O)C(=O)O

Specifications

Linear Formula C19H35N.C4H4O4
Quantity 50 mg
Solubility Information Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 393.56
Chemical Name or Material 2-(2,2-Dicyclohexylethyl)piperidine maleate
Grade >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.