Learn More
Cytiva Peristaltic Pump
Single-channel peristaltic pump combines reproducible flow rates with simple operation. Cytiva Peristaltic Pump functions either as a standalone unit, or interfaced with programmable fraction collectors.
Brand:  Cytiva 18-1110-91
Product Code. 11340142
  • £2766.50 / Each
Explore more special offers
Description
Description
Features
  • Low pulsation flow from 0.6 to 500 mL/h,
  • Automatic pressure plate for convenient tubing changes
  • Maximum and reverse flow functions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 each silicone tubing 1mm, 2.1mm, 3.1mm i.d., 6 tubing connectors, Allen key 3mm, Flow rate guide, O-ring fuse and fuse, Support rod/P-1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Peristaltic Pump > Type: Peristaltic