missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow 120 Series Drive with Pumphead

120 series pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.

Brand:  Watson-Marlow 010.6131.M30

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12446260

  • £5235.00 / Each
Estimated Shipment: 30-08-2024
to see stock.

Description

Description

  • The 120 series is the perfect pump for maximising yield in fermentation and cell culture applications. Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimise your process. The 120 series pump is perfectly suited to single-use systems. Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate. With an exceptional 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials. A keypad lock secures your pumping parameters for consistent and repeatable performance.
  • Pumphead options available as standard (others available on request):

  • 120R single channel, two roller pumphead for continuous tubing
Specifications

Specifications

Digital Variable Speed
IP31
30 psi
120S/DM3
90/264 V
LED
0.0001 to 36 mL/min.
0.01 to 100 rpm
400DM3 pump head
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers