Learn More
Watson-Marlow 120 Series Drive with Pumphead
120 series pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.
Brand:  Watson-Marlow 010.6141.DA0
3135.29 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 12476260
  • £6375.00 / Each
Explore more special offers
Description
Description
  • The 120 series is the perfect pump for maximising yield in fermentation and cell culture applications. Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimise your process. The 120 series pump is perfectly suited to single-use systems. Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate. With an exceptional 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials. A keypad lock secures your pumping parameters for consistent and repeatable performance.
  • Pumphead options available as standard (others available on request):
  • 120R single channel, two roller pumphead for continuous tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital Variable Speed
IP31
30 psi
120U/DV
90/264 V
LED
0.002 to 190 mL/min.
0.1 to 200 rpm
114DV flip-top pump head