Learn More
PERK Monoclonal Antibody (5G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515705
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15705 targets EIF2AK3 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15705 immunogen is purified recombinant fragment of human EIF2AK3 expressed in E. Coli. MA5-15705 detects EIF2AK3 which has a predicted molecular weight of approximately 125kDa.
EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.Specifications
PERK | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9NZJ5 | |
EIF2AK3 | |
Purified recombinant fragment of human EIF2AK3 expressed in E. Coli. | |
RUO | |
9451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5G5 | |
Unconjugated | |
EIF2AK3 | |
2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |